The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164840.2 | NP_001158312.1 | 1 - 2 | 296 | 69 |
NM_001164841.2 | NP_001158313.1 | 1 - 2 | 296 | 69 | |
NM_001318782.1 | NP_001305711.1 | 1 - 2 | 296 | 69 | |
NM_001318783.1 | NP_001305712.1 | 1 - 2 | 296 | 69 | |
NM_020408.5 | NP_065141.3 | 1 - 2 | 296 | 69 | |
NR_104417.1 | 1 - 2 | 296 | 69 | ||
NR_134856.1 | 1 - 2 | 296 | 69 | ||
XM_017011083.1 | XP_016866572.1 | 1 - 2 | 290 | 69 | |
XM_017011084.1 | XP_016866573.1 | 1 - 2 | 293 | 69 | |
GenBank mRNA | AF088060.1 | - | 1 - 2 | 76 | 69 |
AF170070.1 | - | 1 - 2 | 92 | 69 | |
AF285118.1 | - | 1 - 2 | 92 | 69 | |
AK291158.1 | - | 1 - 2 | 88 | 69 | |
AK309661.1 | - | 1 - 2 | 98 | 69 | |
BC009552.2 | - | 1 - 2 | 147 | 69 | |
BI226933.1 | - | 1 - 2 | 270 | 69 | |
BI753819.1 | - | 1 - 2 | 282 | 69 | |
BI758258.1 | - | 1 - 2 | 285 | 69 | |
BX353584.2 | - | 1 - 2 | 75 | 69 | |
CK819966.1 | - | 1 - 2 | 72 | 69 | |
DA114665.1 | - | 1 - 2 | 88 | 69 | |
DA222333.1 | - | 1 - 2 | 88 | 69 | |
HY168919.1 | - | 1 - 2 | 297 | 69 |