The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306135.1 | NP_001293064.1 | 1 - 2 | 746 | 131 |
GenBank mRNA | AK303567.1 | - | 2 - 3 | 563 | 131 |
AY357595.1 | - | 1 - 2 | 503 | 131 | |
DN992279.1 | - | 1 - 2 | 329 | 131 |