The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134445.1 | NP_001127917.1 | 1 - 2 | 376 | 71 |
XM_011541158.1 | XP_011539460.1 | 1 - 2 | 287 | 71 | |
GenBank mRNA | AK304876.1 | - | 1 - 2 | 380 | 71 |
BX648145.1 | - | 1 - 2 | 263 | 71 |