The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178536.3 | NP_848631.2 | 1 - 2 | 124 | 56 |
XM_006717065.3 | XP_006717128.1 | 2 - 3 | 609 | 56 | |
XM_011518560.2 | XP_011516862.1 | 2 - 3 | 761 | 56 | |
XM_011518561.2 | XP_011516863.2 | 1 - 2 | 301 | 56 | |
XM_011518562.2 | XP_011516864.1 | 2 - 3 | 162 | 56 | |
XM_017014631.1 | XP_016870120.1 | 1 - 2 | 300 | 56 | |
XM_017014632.1 | XP_016870121.1 | 1 - 2 | 296 | 56 | |
GenBank mRNA | AY301272.1 | - | 1 - 2 | 112 | 56 |
BC041168.1 | - | 1 - 2 | 124 | 56 | |
BX281250.1 | - | 1 - 2 | 124 | 56 |