The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080436.1 | NP_001073905.1 | 4 - 5 | 906 | 65 |
XM_006723014.3 | XP_006723077.1 | 4 - 5 | 939 | 65 | |
XM_011526452.2 | XP_011524754.1 | 4 - 5 | 946 | 65 | |
XM_011526453.2 | XP_011524755.1 | 4 - 5 | 340 | 65 | |
GenBank mRNA | AK130059.1 | - | 3 - 4 | 244 | 65 |