Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ECSCR
Entrez Gene ID
641700
Gene Name
endothelial cell surface expressed chemotaxis and apoptosis regulator
Gene Aliases
ARIA, ECSM2
Chromosome Location
Chr.5: 139448554 - 139462743 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077693.3 NP_001071161.1 9 - 10 683 117
GenBank mRNA BC146912.1 - 9 - 10 655 117
BC146913.1 - 9 - 10 655 117
DQ462572.1 - 9 - 10 677 117
EU025066.1 - 9 - 10 683 117

View Full Product Details