Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- OVOS2
- Entrez Gene ID
-
144203
- Gene Name
- ovostatin 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.12: 31114623 - 31206154 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AL831947.1 |
- |
- |
2158 |
125 |
BC039117.1 |
- |
4 - 5 |
1033 |
125 |
BC046190.1 |
- |
3 - 4 |
382 |
125 |
CR749395.1 |
- |
3 - 4 |
2346 |
125 |
- Gene Symbol
- OVOS
- Entrez Gene ID
-
408186
- Gene Name
- ovostatin
- Gene Aliases
-
-
- Chromosome Location
-
Chr.12: 9517803 - 9573287 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BX647938.1 |
- |
10 - 11 |
6375 |
125 |
View Full Product Details