The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002254.1 | NP_001002254.1 | 3 - 4 | 268 | 94 |
XM_011530876.2 | XP_011529178.1 | 3 - 4 | 300 | 94 | |
GenBank mRNA | AY605053.1 | - | 3 - 4 | 268 | 94 |