The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001123225.1 | NP_001116697.1 | 2 - 3 | 152 | 86 |
GenBank mRNA | BC127859.1 | - | 1 - 2 | 137 | 86 |
BC127860.1 | - | 1 - 2 | 137 | 86 | |
EU082028.1 | - | 1 - 2 | 110 | 86 |