The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004168.3 | NP_004159.2 | 4 - 5 | 587 | 124 |
XM_005248331.3 | XP_005248388.1 | 4 - 5 | 587 | 124 | |
XM_011514072.1 | XP_011512374.1 | 4 - 5 | 587 | 124 | |
XM_011514073.1 | XP_011512375.1 | 4 - 5 | 587 | 124 | |
XM_017009685.1 | XP_016865174.1 | 4 - 5 | 587 | 124 | |
GenBank mRNA | AB208991.1 | - | 4 - 5 | 473 | 124 |
AK225027.1 | - | 4 - 5 | 488 | 124 | |
AK291311.1 | - | 4 - 5 | 486 | 124 | |
AK302524.1 | - | 4 - 5 | 494 | 124 | |
BC001380.2 | - | 4 - 5 | 483 | 124 | |
D30648.1 | - | 4 - 5 | 460 | 124 | |
DQ402982.1 | - | 2 - 3 | 259 | 124 | |
L21936.1 | - | 4 - 5 | 478 | 124 |