Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAH14
Entrez Gene ID
127602
Gene Name
dynein axonemal heavy chain 14
Gene Aliases
C1orf67, Dnahc14, HL-18, HL18
Chromosome Location
Chr.1: 224928422 - 225399294 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373.1 NP_001364.1 49 - 50 7734 112
XM_006711735.3 XP_006711798.1 49 - 50 8983 112
XM_011544057.2 XP_011542359.1 49 - 50 8983 112
XM_011544058.2 XP_011542360.1 49 - 50 8980 112
XM_011544059.2 XP_011542361.1 49 - 50 8935 112
XM_011544061.2 XP_011542363.1 49 - 50 8878 112
XM_011544062.2 XP_011542364.1 49 - 50 8983 112
XM_011544063.2 XP_011542365.1 48 - 49 8824 112
XM_011544064.2 XP_011542366.1 49 - 50 8983 112
XM_011544065.2 XP_011542367.1 49 - 50 8983 112
XM_011544066.2 XP_011542368.1 48 - 49 8767 112
XM_011544067.2 XP_011542369.1 49 - 50 8983 112
XM_011544069.2 XP_011542371.1 49 - 50 8983 112
XM_011544070.2 XP_011542372.1 47 - 48 8605 112
XM_011544071.2 XP_011542373.1 46 - 47 8456 112
XM_011544072.2 XP_011542374.1 42 - 43 7666 112
XM_011544073.2 XP_011542375.1 36 - 37 8599 112
XM_011544074.2 XP_011542376.1 33 - 34 5836 112
XM_011544076.1 XP_011542378.1 31 - 32 4488 112
XM_011544079.1 XP_011542381.1 24 - 25 3221 112
XM_011544081.1 XP_011542383.1 15 - 16 2171 112
XM_017000295.1 XP_016855784.1 48 - 49 8908 112
XM_017000296.1 XP_016855785.1 49 - 50 8983 112
XM_017000297.1 XP_016855786.1 48 - 49 8866 112
XM_017000298.1 XP_016855787.1 49 - 50 8982 112
XM_017000299.1 XP_016855788.1 23 - 24 3067 112
XM_017000300.1 XP_016855789.1 49 - 50 8977 112
GenBank mRNA CR936804.1 - 7 - 8 818 112

View Full Product Details