Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C3P1
Entrez Gene ID
388503
Gene Name
complement component 3 precursor pseudogene
Gene Aliases
CPLP
Chromosome Location
Chr.19: 10041356 - 10074138 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_027300.2 28 - 29 3413 78
GenBank mRNA AK131489.1 - 26 - 27 3178 78

View Full Product Details