Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BTBD17
Entrez Gene ID
388419
Gene Name
BTB domain containing 17
Gene Aliases
BTBD17A, LGALS3BPL, TANGO10A
Chromosome Location
Chr.17: 74356416 - 74363655 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080466.1 NP_001073935.1 2 - 3 358 54
XM_011524791.2 XP_011523093.1 4 - 5 1264 54
XM_017024622.1 XP_016880111.1 4 - 5 1200 54

View Full Product Details