Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OTUD3
Entrez Gene ID
23252
Gene Name
OTU deubiquitinase 3
Gene Aliases
DUBA4
Chromosome Location
Chr.1: 19881711 - 19912945 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015207.1 NP_056022.1 3 - 4 597 89
XM_005245792.3 XP_005245849.1 3 - 4 1280 89
XM_005245793.3 XP_005245850.1 3 - 4 1281 89
GenBank mRNA AB007928.3 - 3 - 4 574 89

View Full Product Details