Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VSTM5
Entrez Gene ID
387804
Gene Name
V-set and transmembrane domain containing 5
Gene Aliases
C11orf90
Chromosome Location
Chr.11: 93817315 - 93850653 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144871.1 NP_001138343.1 3 - 4 566 106

View Full Product Details