The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207363.2 | NP_997246.2 | 18 - 19 | 5953 | 84 |
NM_207481.3 | NP_997364.3 | 16 - 17 | 1996 | 84 | |
XM_017003980.1 | XP_016859469.1 | 13 - 14 | 1897 | 84 | |
GenBank mRNA | AK124659.1 | - | 5 - 6 | 825 | 84 |
AY946007.1 | - | 11 - 12 | 1418 | 84 | |
AY946008.1 | - | 13 - 14 | 5375 | 84 |