Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TTC24
Entrez Gene ID
164118
Gene Name
tetratricopeptide repeat domain 24
Gene Aliases
-
Chromosome Location
Chr.1: 156579727 - 156587717 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105669.2 NP_001099139.2 5 - 6 1186 81
XM_011509253.2 XP_011507555.1 4 - 5 568 81
XM_011509255.2 XP_011507557.1 4 - 5 568 81
XM_017000497.1 XP_016855986.1 2 - 3 190 81
GenBank mRNA BQ216668.1 - 3 - 4 450 81

View Full Product Details