The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005751.2 | NP_001005751.1 | 20 - 21 | 2155 | 119 |
NM_001291398.1 | NP_001278327.1 | 20 - 21 | 2155 | 119 | |
XM_005269806.3 | XP_005269863.2 | 19 - 20 | 2031 | 119 | |
XM_005269807.3 | XP_005269864.2 | 19 - 20 | 2016 | 119 | |
XM_005269808.3 | XP_005269865.2 | 20 - 21 | 2103 | 119 | |
XM_006717831.3 | XP_006717894.2 | 20 - 21 | 2103 | 119 | |
XM_006717832.3 | XP_006717895.2 | 19 - 20 | 2031 | 119 | |
XM_011539786.2 | XP_011538088.1 | 20 - 21 | 2103 | 119 | |
XM_017016227.1 | XP_016871716.1 | 20 - 21 | 2103 | 119 | |
XM_017016228.1 | XP_016871717.1 | 19 - 20 | 2031 | 119 | |
XM_017016229.1 | XP_016871718.1 | 19 - 20 | 2016 | 119 | |
XM_017016230.1 | XP_016871719.1 | 19 - 20 | 1968 | 119 | |
XM_017016231.1 | XP_016871720.1 | 20 - 21 | 2103 | 119 | |
XM_017016232.1 | XP_016871721.1 | 18 - 19 | 1944 | 119 | |
XM_017016233.1 | XP_016871722.1 | 17 - 18 | 1678 | 119 | |
XM_017016234.1 | XP_016871723.1 | 19 - 20 | 2038 | 119 | |
GenBank mRNA | AK001686.1 | - | 17 - 18 | 1807 | 119 |
AK295129.1 | - | 20 - 21 | 2107 | 119 | |
AK304116.1 | - | 20 - 21 | 2091 | 119 | |
BC075815.1 | - | 17 - 18 | 1720 | 119 | |
BC082258.1 | - | 20 - 21 | 2082 | 119 | |
BC144479.1 | - | 20 - 21 | 2184 | 119 | |
BE744488.1 | - | 5 - 6 | 549 | 119 | |
DA466642.1 | - | 2 - 3 | 306 | 119 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001169106.1 | NP_001162577.1 | 20 - 21 | 2155 | 119 |
NM_001169107.1 | NP_001162578.1 | 19 - 20 | 2083 | 119 | |
NM_015262.2 | NP_056077.2 | 20 - 21 | 2155 | 119 | |
XM_011539566.1 | XP_011537868.1 | 20 - 21 | 2122 | 119 | |
XM_011539567.1 | XP_011537869.1 | 19 - 20 | 2050 | 119 | |
XM_011539568.1 | XP_011537870.1 | 19 - 20 | 2035 | 119 | |
XM_011539569.1 | XP_011537871.1 | 20 - 21 | 2122 | 119 | |
XM_011539570.2 | XP_011537872.1 | 19 - 20 | 2011 | 119 | |
XM_011539571.1 | XP_011537873.1 | 20 - 21 | 2122 | 119 | |
XM_017016014.1 | XP_016871503.1 | 20 - 21 | 2122 | 119 | |
XM_017016015.1 | XP_016871504.1 | 19 - 20 | 2050 | 119 | |
XM_017016016.1 | XP_016871505.1 | 19 - 20 | 2035 | 119 | |
XM_017016017.1 | XP_016871506.1 | 18 - 19 | 1963 | 119 | |
XM_017016018.1 | XP_016871507.1 | 20 - 21 | 2122 | 119 | |
XM_017016019.1 | XP_016871508.1 | 19 - 20 | 2050 | 119 | |
XM_017016020.1 | XP_016871509.1 | 19 - 20 | 2050 | 119 | |
XM_017016021.1 | XP_016871510.1 | 20 - 21 | 2050 | 119 | |
XM_017016022.1 | XP_016871511.1 | 20 - 21 | 2096 | 119 | |
XM_017016023.1 | XP_016871512.1 | 19 - 20 | 1968 | 119 | |
GenBank mRNA | AB011164.1 | - | 20 - 21 | 2136 | 119 |
AK001545.1 | - | 2 - 3 | 316 | 119 | |
AK291430.1 | - | 20 - 21 | 2143 | 119 | |
AK293929.1 | - | 15 - 16 | 1606 | 119 | |
AK298385.1 | - | 18 - 19 | 1820 | 119 | |
AK303048.1 | - | 19 - 20 | 2047 | 119 | |
BC053887.1 | - | 19 - 20 | 1901 | 119 | |
BC150611.1 | - | 20 - 21 | 2120 | 119 |