Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNX6
Entrez Gene ID
58533
Gene Name
sorting nexin 6
Gene Aliases
MSTP010, TFAF2
Chromosome Location
Chr.14: 34561409 - 34630160 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK296213.1 - 6 - 7 563 94
AK308464.1 - 6 - 7 563 94

View Full Product Details