The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153046.2 | NP_694591.2 | 4 - 5 | 667 | 91 |
XM_005267309.3 | XP_005267366.1 | 4 - 5 | 804 | 91 | |
XM_006720019.3 | XP_006720082.1 | 4 - 5 | 804 | 91 | |
XM_006720020.3 | XP_006720083.1 | 4 - 5 | 804 | 91 | |
XM_011536397.2 | XP_011534699.1 | 4 - 5 | 804 | 91 | |
XM_011536398.2 | XP_011534700.1 | 4 - 5 | 609 | 91 | |
XM_011536400.2 | XP_011534702.1 | 4 - 5 | 804 | 91 | |
XM_011536401.2 | XP_011534703.1 | 4 - 5 | 542 | 91 | |
XM_017020955.1 | XP_016876444.1 | 4 - 5 | 658 | 91 | |
GenBank mRNA | AL833915.1 | - | 1 - 2 | 57 | 91 |
U56244.1 | - | 3 - 4 | 406 | 91 |