The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001115007.2 | NP_001108479.1 | 10 - 11 | 1125 | 88 |
NM_001115008.2 | NP_001108480.1 | 10 - 11 | 1326 | 88 | |
NM_001288996.1 | NP_001275925.1 | 7 - 8 | 1722 | 88 | |
NM_001288997.1 | NP_001275926.1 | 10 - 11 | 1063 | 88 | |
NM_194282.3 | NP_919258.2 | 10 - 11 | 1989 | 88 | |
NR_110253.1 | 7 - 8 | 1648 | 88 | ||
NR_110254.1 | 9 - 10 | 1322 | 88 | ||
XM_005262750.4 | XP_005262807.1 | 9 - 10 | 1798 | 88 | |
XM_006714081.3 | XP_006714144.1 | 9 - 10 | 1724 | 88 | |
XM_017007728.1 | XP_016863217.1 | 7 - 8 | 1531 | 88 | |
XM_017007729.1 | XP_016863218.1 | 8 - 9 | 1714 | 88 | |
XM_017007730.1 | XP_016863219.1 | 8 - 9 | 1103 | 88 | |
GenBank mRNA | AB111889.1 | - | 9 - 10 | 1309 | 88 |
AK292769.1 | - | 10 - 11 | 1073 | 88 | |
AK294256.1 | - | 10 - 11 | 1918 | 88 | |
AK299787.1 | - | 5 - 6 | 531 | 88 | |
BC109277.2 | - | 7 - 8 | 1524 | 88 | |
BC109278.2 | - | 7 - 8 | 1597 | 88 | |
BX537919.1 | - | 9 - 10 | 1223 | 88 | |
BX640657.1 | - | 10 - 11 | 1052 | 88 | |
BX640966.1 | - | 10 - 11 | 1984 | 88 |