The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_206998.1 | NP_996881.1 | 1 - 2 | 113 | 70 |
GenBank mRNA | AY236538.1 | - | 1 - 2 | 80 | 70 |
AY359052.1 | - | 1 - 2 | 113 | 70 | |
BC130639.1 | - | 1 - 2 | 88 | 70 | |
BC130645.1 | - | 1 - 2 | 88 | 70 |