The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113523.2 | NP_001106995.1 | 2 - 3 | 374 | 85 |
NM_001308320.1 | NP_001295249.1 | 2 - 3 | 374 | 85 | |
XM_005247159.3 | XP_005247216.1 | 2 - 3 | 2046 | 85 | |
XM_005247160.3 | XP_005247217.1 | 2 - 3 | 2046 | 85 | |
XM_011512476.2 | XP_011510778.1 | 2 - 3 | 2049 | 85 | |
XM_011512477.2 | XP_011510779.1 | 2 - 3 | 2046 | 85 | |
XM_011512478.2 | XP_011510780.1 | 1 - 2 | 122 | 85 | |
XM_011512480.2 | XP_011510782.1 | 2 - 3 | 2062 | 85 | |
XM_017005791.1 | XP_016861280.1 | 2 - 3 | 2046 | 85 | |
GenBank mRNA | AK097515.1 | - | 3 - 4 | 284 | 85 |
AK309863.1 | - | 2 - 3 | 374 | 85 | |
BX647656.1 | - | 2 - 3 | 350 | 85 | |
DQ063586.1 | - | 2 - 3 | 293 | 85 |