The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_017008241.1 | XP_016863730.1 | 3 - 4 | 1930 | 110 |
GenBank mRNA | BC127739.1 | - | 1 - 2 | 127 | 110 |
BC141968.1 | - | 1 - 2 | 126 | 110 |