The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010906.1 | NP_001010906.1 | 7 - 8 | 1063 | 72 |
XM_011544523.2 | XP_011542825.1 | 7 - 8 | 1332 | 72 | |
XM_011544524.2 | XP_011542826.1 | 7 - 8 | 2838 | 72 | |
XM_011544526.2 | XP_011542828.1 | 7 - 8 | 1332 | 72 | |
XM_017013403.1 | XP_016868892.1 | 7 - 8 | 1332 | 72 | |
GenBank mRNA | AB075870.1 | - | 7 - 8 | 1064 | 72 |