The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135219.1 | NP_001128691.1 | 1 - 2 | 77 | 101 |
XM_017014279.1 | XP_016869768.1 | 1 - 2 | 1238 | 101 |