Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDX1
Entrez Gene ID
3651
Gene Name
pancreatic and duodenal homeobox 1
Gene Aliases
GSF, IDX-1, IPF1, IUF1, MODY4, PAGEN1, PDX-1, STF-1
Chromosome Location
Chr.13: 27919982 - 27926314 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000209.3 NP_000200.1 1 - 2 512 153
GenBank mRNA U30329.1 - 1 - 2 424 153
U35632.1 - 1 - 2 418 153
X99894.1 - 1 - 2 503 153

View Full Product Details