Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP2C18
Entrez Gene ID
1562
Gene Name
cytochrome P450 family 2 subfamily C member 18
Gene Aliases
CPCI, CYP2C, CYP2C17, P450-6B/29C, P450IIC17
Chromosome Location
Chr.10: 94683494 - 94736190 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000772.2 NP_000763.1 1 - 2 490 94
NM_001128925.1 NP_001122397.1 1 - 2 490 94
GenBank mRNA AK223510.1 - 1 - 2 189 94
AK313403.1 - 1 - 2 189 94
BC069666.1 - 1 - 2 358 94
BC096257.1 - 1 - 2 209 94
BC096258.1 - 1 - 2 209 94
BC096260.1 - 1 - 2 209 94
M61853.1 - 1 - 2 206 94
M61856.1 - 1 - 2 363 94

View Full Product Details