The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172085.1 | NP_001165556.1 | 3 - 4 | 666 | 65 |
NM_003194.4 | NP_003185.1 | 4 - 5 | 868 | 65 | |
GenBank mRNA | AK298272.1 | - | 5 - 6 | 750 | 65 |
AK304648.1 | - | 3 - 4 | 653 | 65 | |
AK315222.1 | - | 4 - 5 | 831 | 65 | |
BC109053.1 | - | 4 - 5 | 779 | 65 | |
BC109054.1 | - | 5 - 6 | 633 | 65 | |
BC110341.1 | - | 4 - 5 | 827 | 65 | |
BT019657.1 | - | 3 - 4 | 580 | 65 | |
CR456776.1 | - | 3 - 4 | 589 | 65 | |
M55654.1 | - | 4 - 5 | 830 | 65 | |
X54993.1 | - | 3 - 4 | 675 | 65 |