The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001265591.1 | NP_001252520.1 | 1 - 2 | 326 | 154 |
NM_006054.3 | NP_006045.1 | 1 - 2 | 326 | 154 | |
NM_201430.2 | NP_958833.1 | 1 - 2 | 326 | 154 | |
NR_049750.1 | 1 - 2 | 326 | 154 | ||
NR_049751.1 | 1 - 2 | 326 | 154 | ||
GenBank mRNA | AB209771.1 | - | 1 - 2 | 263 | 154 |
AF059524.1 | - | 1 - 2 | 263 | 154 | |
AF119297.1 | - | 1 - 2 | 275 | 154 | |
AK075412.1 | - | 1 - 2 | 276 | 154 | |
AK222898.1 | - | 1 - 2 | 263 | 154 | |
AK297529.1 | - | 1 - 2 | 203 | 154 | |
AK297533.1 | - | 1 - 2 | 276 | 154 | |
AK308083.1 | - | 1 - 2 | 271 | 154 | |
BC000634.1 | - | 1 - 2 | 234 | 154 | |
BC010556.1 | - | 1 - 2 | 239 | 154 | |
BC011394.1 | - | 1 - 2 | 275 | 154 | |
BC022993.1 | - | 1 - 2 | 265 | 154 | |
BC100822.2 | - | 1 - 2 | 256 | 154 | |
BC100823.2 | - | 1 - 2 | 256 | 154 | |
BC105981.1 | - | 1 - 2 | 207 | 154 | |
BC105982.1 | - | 1 - 2 | 238 | 154 | |
BC118550.1 | - | 1 - 2 | 256 | 154 | |
BC118628.1 | - | 1 - 2 | 256 | 154 |