Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAN2C1
Entrez Gene ID
4123
Gene Name
mannosidase alpha class 2C member 1
Gene Aliases
MAN6A8, MANA, MANA1
Chromosome Location
Chr.15: 75355792 - 75368680 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256494.1 NP_001243423.1 9 - 10 1138 86
NM_001256495.1 NP_001243424.1 9 - 10 1138 86
NM_001256496.1 NP_001243425.1 7 - 8 841 86
NM_006715.3 NP_006706.2 9 - 10 1138 86
XM_005254384.2 XP_005254441.1 9 - 10 1191 86
XM_017022186.1 XP_016877675.1 8 - 9 1486 86
XM_017022187.1 XP_016877676.1 8 - 9 1486 86
XM_017022188.1 XP_016877677.1 7 - 8 894 86
GenBank mRNA AB209141.1 - - 2455 86
AF044414.2 - 9 - 10 1150 86
AK225145.1 - 7 - 8 818 86
AK294892.1 - 8 - 9 927 86
AK311065.1 - 8 - 9 990 86
AL136876.1 - 9 - 10 1099 86
BC050550.1 - 9 - 10 1105 86
BC080191.1 - 9 - 10 1103 86

View Full Product Details