The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015995.3 | NP_057079.2 | 1 - 2 | 941 | 107 |
GenBank mRNA | AF132599.1 | - | 1 - 2 | 955 | 107 |
AF150628.1 | - | 1 - 2 | 634 | 107 | |
BC010438.1 | - | 1 - 2 | 896 | 107 | |
BC012741.2 | - | 1 - 2 | 799 | 107 | |
BC013946.2 | - | 1 - 2 | 896 | 107 | |
BC035503.2 | - | 1 - 2 | 916 | 107 | |
KJ534876.1 | - | 1 - 2 | 583 | 107 | |
KJ535015.1 | - | 2 - 3 | 383 | 107 |