Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 24123688-24124185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRKCB
Entrez Gene ID
5579
Gene Name
protein kinase C beta
Gene Aliases
PKC-beta, PKCB, PRKCB1, PRKCB2
UniGene
Hs.460355
Gene Chromosome Location
Chr.16: 23835979-24220611 on Build GRCh38
Transcripts
NM_002738.6 NM_212535.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7193043 c.1016T>C p.L339P Chr.16: 24123932-24123932 on Build GRCh38
1581529 c.1055G>A p.S352N Chr.16: 24123971-24123971 on Build GRCh38
1222019 c.950C>T p.P317L Chr.16: 24123866-24123866 on Build GRCh38
6859400 c.1006C>T p.R336W Chr.16: 24123922-24123922 on Build GRCh38
1263066 c.1007G>A p.R336Q Chr.16: 24123923-24123923 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs114856527 Chr.16: 24123897-24123897 on Build GRCh38
rs757243180 Chr.16: 24124008-24124008 on Build GRCh38
rs539605078 Chr.16: 24124080-24124080 on Build GRCh38
rs895954203 Chr.16: 24123749-24123749 on Build GRCh38
rs1015827733 Chr.16: 24124001-24124001 on Build GRCh38
rs934703106 Chr.16: 24124061-24124061 on Build GRCh38
rs749243156 Chr.16: 24123896-24123896 on Build GRCh38
rs866640642 Chr.16: 24123854-24123854 on Build GRCh38
rs753779451 Chr.16: 24124004-24124004 on Build GRCh38
rs1005918505 Chr.16: 24124148-24124148 on Build GRCh38
rs751175487 Chr.16: 24123880-24123880 on Build GRCh38
rs766920124 Chr.16: 24123955-24123955 on Build GRCh38
rs750938251 Chr.16: 24123867-24123867 on Build GRCh38
rs551131907 Chr.16: 24123735-24123735 on Build GRCh38
rs780590117 Chr.16: 24123799-24123799 on Build GRCh38
rs768249184 Chr.16: 24123821-24123822 on Build GRCh38
rs1013316 Chr.16: 24123791-24123791 on Build GRCh38
rs116346810 Chr.16: 24123936-24123936 on Build GRCh38
rs202044032 Chr.16: 24123866-24123866 on Build GRCh38
rs550804620 Chr.16: 24124147-24124147 on Build GRCh38
rs149128755 Chr.16: 24123911-24123911 on Build GRCh38
rs918034435 Chr.16: 24124094-24124094 on Build GRCh38
rs896039726 Chr.16: 24124089-24124089 on Build GRCh38
rs949382713 Chr.16: 24124103-24124103 on Build GRCh38
rs78246203 Chr.16: 24123721-24123721 on Build GRCh38
rs1043195237 Chr.16: 24124048-24124048 on Build GRCh38
rs374448988 Chr.16: 24123809-24123809 on Build GRCh38
rs185274925 Chr.16: 24124072-24124072 on Build GRCh38
rs36096722 Chr.16: 24123995-24123996 on Build GRCh38
rs1013592826 Chr.16: 24124118-24124118 on Build GRCh38
rs763468825 Chr.16: 24123954-24123954 on Build GRCh38
rs190305376 Chr.16: 24124083-24124083 on Build GRCh38
rs779621369 Chr.16: 24123789-24123789 on Build GRCh38
rs781201941 Chr.16: 24124029-24124029 on Build GRCh38
rs748429648 Chr.16: 24124031-24124031 on Build GRCh38
rs772280177 Chr.16: 24124127-24124127 on Build GRCh38
rs749777388 Chr.16: 24123862-24123862 on Build GRCh38
rs778877195 Chr.16: 24123906-24123906 on Build GRCh38
rs771911978 Chr.16: 24124018-24124018 on Build GRCh38
rs539838175 Chr.16: 24123924-24123924 on Build GRCh38
rs201781724 Chr.16: 24123846-24123846 on Build GRCh38
rs756613827 Chr.16: 24123733-24123733 on Build GRCh38
rs767062481 Chr.16: 24123985-24123985 on Build GRCh38
rs755619557 Chr.16: 24123993-24123993 on Build GRCh38
rs748379616 Chr.16: 24124013-24124013 on Build GRCh38
rs755710261 Chr.16: 24123893-24123893 on Build GRCh38
rs777796805 Chr.16: 24123894-24123894 on Build GRCh38
rs758818157 Chr.16: 24123786-24123786 on Build GRCh38
rs146104501 Chr.16: 24123861-24123861 on Build GRCh38
rs201114095 Chr.16: 24123817-24123817 on Build GRCh38
rs1023102067 Chr.16: 24123790-24123790 on Build GRCh38
rs561867907 Chr.16: 24124038-24124038 on Build GRCh38
rs768732945 Chr.16: 24123922-24123922 on Build GRCh38
rs903169006 Chr.16: 24124060-24124060 on Build GRCh38
rs747742173 Chr.16: 24123805-24123805 on Build GRCh38
rs1027953347 Chr.16: 24123918-24123918 on Build GRCh38
rs754466595 Chr.16: 24123793-24123793 on Build GRCh38
rs535322667 Chr.16: 24124075-24124076 on Build GRCh38
rs749083935 Chr.16: 24123818-24123818 on Build GRCh38
rs769585977 Chr.16: 24123807-24123807 on Build GRCh38
rs576546931 Chr.16: 24124064-24124064 on Build GRCh38
rs777457442 Chr.16: 24124000-24124000 on Build GRCh38
rs752410072 Chr.16: 24123992-24123992 on Build GRCh38
rs760838283 Chr.16: 24123839-24123839 on Build GRCh38
rs775788179 Chr.16: 24123829-24123829 on Build GRCh38
rs542202519 Chr.16: 24124165-24124165 on Build GRCh38
rs776981512 Chr.16: 24123934-24123934 on Build GRCh38
rs766756709 Chr.16: 24123879-24123879 on Build GRCh38
rs569847148 Chr.16: 24123769-24123769 on Build GRCh38
rs374744056 Chr.16: 24123891-24123891 on Build GRCh38
rs770864109 Chr.16: 24123825-24123825 on Build GRCh38
rs773665394 Chr.16: 24123952-24123952 on Build GRCh38
rs139055681 Chr.16: 24123878-24123878 on Build GRCh38
rs141851225 Chr.16: 24123836-24123836 on Build GRCh38
rs754563290 Chr.16: 24123881-24123881 on Build GRCh38
rs990869257 Chr.16: 24124075-24124075 on Build GRCh38
rs764189258 Chr.16: 24123844-24123844 on Build GRCh38
rs745649033 Chr.16: 24124009-24124009 on Build GRCh38
rs56041613 Chr.16: 24123945-24123945 on Build GRCh38
rs556610239 Chr.16: 24123978-24123978 on Build GRCh38
rs780936454 Chr.16: 24123888-24123888 on Build GRCh38

View Full Product Details