Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43605114-43605690 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLH
Entrez Gene ID
5429
Gene Name
DNA polymerase eta
Gene Aliases
RAD30, RAD30A, XP-V, XPV
UniGene
Hs.655467
Gene Chromosome Location
Chr.6: 43576141-43620523 on Build GRCh38
Transcripts
NM_001291969.1 NM_001291970.1 NM_006502.2 XM_005249186.3 XM_011514698.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7285339 c.1009-8delT p.? Chr.6: 43605246-43605246 on Build GRCh38
1079647 c.1067G>A p.R356Q Chr.6: 43605312-43605312 on Build GRCh38
1079646 c.1030T>A p.L344I Chr.6: 43605275-43605275 on Build GRCh38
5782063 c.1072G>A p.D358N Chr.6: 43605317-43605317 on Build GRCh38
6752101 c.1066C>T p.R356* Chr.6: 43605311-43605311 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs878858171 Chr.6: 43605467-43605468 on Build GRCh38
rs13197957 Chr.6: 43605444-43605444 on Build GRCh38
rs1000673974 Chr.6: 43605464-43605464 on Build GRCh38
rs548895303 Chr.6: 43605480-43605480 on Build GRCh38
rs200775544 Chr.6: 43605269-43605269 on Build GRCh38
rs756481594 Chr.6: 43605433-43605433 on Build GRCh38
rs1031273064 Chr.6: 43605488-43605488 on Build GRCh38
rs904611245 Chr.6: 43605452-43605452 on Build GRCh38
rs369142296 Chr.6: 43605212-43605212 on Build GRCh38
rs759317236 Chr.6: 43605597-43605597 on Build GRCh38
rs948847190 Chr.6: 43605286-43605286 on Build GRCh38
rs9333551 Chr.6: 43605347-43605347 on Build GRCh38
rs755763217 Chr.6: 43605323-43605323 on Build GRCh38
rs887113551 Chr.6: 43605341-43605341 on Build GRCh38
rs754126851 Chr.6: 43605196-43605196 on Build GRCh38
rs1024590106 Chr.6: 43605541-43605541 on Build GRCh38
rs543410206 Chr.6: 43605169-43605169 on Build GRCh38
rs571720058 Chr.6: 43605579-43605579 on Build GRCh38
rs573883331 Chr.6: 43605404-43605404 on Build GRCh38
rs775604947 Chr.6: 43605274-43605274 on Build GRCh38
rs368893791 Chr.6: 43605345-43605345 on Build GRCh38
rs569386443 Chr.6: 43605623-43605623 on Build GRCh38
rs540143863 Chr.6: 43605444-43605444 on Build GRCh38
rs114052434 Chr.6: 43605497-43605497 on Build GRCh38
rs73428614 Chr.6: 43605366-43605366 on Build GRCh38
rs151138665 Chr.6: 43605574-43605574 on Build GRCh38
rs997711213 Chr.6: 43605581-43605581 on Build GRCh38
rs776938361 Chr.6: 43605266-43605266 on Build GRCh38
rs140119541 Chr.6: 43605580-43605580 on Build GRCh38
rs9333550 Chr.6: 43605156-43605156 on Build GRCh38
rs559497462 Chr.6: 43605311-43605311 on Build GRCh38
rs376692503 Chr.6: 43605217-43605217 on Build GRCh38
rs980849482 Chr.6: 43605547-43605547 on Build GRCh38
rs200060082 Chr.6: 43605443-43605443 on Build GRCh38
rs776415202 Chr.6: 43605583-43605583 on Build GRCh38
rs370554119 Chr.6: 43605439-43605439 on Build GRCh38
rs757424910 Chr.6: 43605320-43605321 on Build GRCh38
rs75277317 Chr.6: 43605259-43605259 on Build GRCh38
rs541116563 Chr.6: 43605287-43605287 on Build GRCh38
rs774088582 Chr.6: 43605183-43605183 on Build GRCh38
rs889154102 Chr.6: 43605442-43605442 on Build GRCh38
rs778439476 Chr.6: 43605536-43605536 on Build GRCh38
rs1021669860 Chr.6: 43605451-43605451 on Build GRCh38
rs767663227 Chr.6: 43605283-43605283 on Build GRCh38
rs990360604 Chr.6: 43605644-43605644 on Build GRCh38
rs373709805 Chr.6: 43605365-43605365 on Build GRCh38
rs143285001 Chr.6: 43605423-43605423 on Build GRCh38
rs991764138 Chr.6: 43605276-43605276 on Build GRCh38
rs560145056 Chr.6: 43605312-43605312 on Build GRCh38
rs1030355631 Chr.6: 43605190-43605190 on Build GRCh38
rs755531601 Chr.6: 43605204-43605204 on Build GRCh38
rs376447275 Chr.6: 43605330-43605330 on Build GRCh38
rs898049032 Chr.6: 43605418-43605418 on Build GRCh38
rs747378313 Chr.6: 43605249-43605249 on Build GRCh38
rs528507039 Chr.6: 43605551-43605551 on Build GRCh38
rs933550440 Chr.6: 43605582-43605582 on Build GRCh38
rs542603998 Chr.6: 43605517-43605517 on Build GRCh38
rs781470373 Chr.6: 43605261-43605261 on Build GRCh38
rs765856970 Chr.6: 43605326-43605326 on Build GRCh38
rs963471848 Chr.6: 43605562-43605562 on Build GRCh38
rs1004208375 Chr.6: 43605439-43605439 on Build GRCh38
rs757650059 Chr.6: 43605368-43605368 on Build GRCh38
rs530816660 Chr.6: 43605465-43605465 on Build GRCh38
rs55737076 Chr.6: 43605447-43605447 on Build GRCh38
rs969066726 Chr.6: 43605546-43605546 on Build GRCh38
rs1048123340 Chr.6: 43605444-43605457 on Build GRCh38
rs12212631 Chr.6: 43605213-43605213 on Build GRCh38
rs557549769 Chr.6: 43605615-43605615 on Build GRCh38
rs778343504 Chr.6: 43605238-43605238 on Build GRCh38
rs71999260 Chr.6: 43605447-43605448 on Build GRCh38
rs774198236 Chr.6: 43605305-43605305 on Build GRCh38
rs374913224 Chr.6: 43605443-43605443 on Build GRCh38
rs751745625 Chr.6: 43605245-43605246 on Build GRCh38
rs910807866 Chr.6: 43605664-43605664 on Build GRCh38
rs886061433 Chr.6: 43605319-43605319 on Build GRCh38

View Full Product Details