Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2597881-2598480 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDPK1
Entrez Gene ID
5170
Gene Name
3-phosphoinositide dependent protein kinase 1
Gene Aliases
PDK1, PDPK2, PDPK2P, PRO0461
UniGene
Hs.459691
Gene Chromosome Location
Chr.16: 2537964-2603190 on Build GRCh38
Transcripts
NM_001261816.1 NM_002613.4 NM_031268.5 XM_011522521.2 XM_011522523.2 XM_017023271.1 XM_017023272.1 XM_017023273.1 XM_017023274.1 XM_017023275.1 XM_017023276.1 XM_017023277.1

SNP Detail

SNP ID Chromosome Location
rs949646710 Chr.16: 2597982-2597982 on Build GRCh38
rs1019429971 Chr.16: 2598028-2598028 on Build GRCh38
rs556257698 Chr.16: 2598353-2598353 on Build GRCh38
rs918073539 Chr.16: 2597970-2597970 on Build GRCh38
rs531570429 Chr.16: 2598146-2598146 on Build GRCh38
rs1024754882 Chr.16: 2598248-2598248 on Build GRCh38
rs925399063 Chr.16: 2597940-2597940 on Build GRCh38
rs908769869 Chr.16: 2598253-2598253 on Build GRCh38
rs901515017 Chr.16: 2597926-2597926 on Build GRCh38
rs767628948 Chr.16: 2597998-2597998 on Build GRCh38
rs1024809062 Chr.16: 2598034-2598034 on Build GRCh38
rs894927700 Chr.16: 2598331-2598331 on Build GRCh38
rs758382220 Chr.16: 2598255-2598255 on Build GRCh38
rs1041233418 Chr.16: 2598097-2598097 on Build GRCh38
rs988213866 Chr.16: 2598064-2598064 on Build GRCh38
rs973120657 Chr.16: 2598447-2598447 on Build GRCh38
rs1047454531 Chr.16: 2598134-2598134 on Build GRCh38
rs935833431 Chr.16: 2598440-2598440 on Build GRCh38
rs1032756249 Chr.16: 2598056-2598056 on Build GRCh38
rs140982354 Chr.16: 2598063-2598063 on Build GRCh38
rs931548413 Chr.16: 2598112-2598112 on Build GRCh38
rs866843949 Chr.16: 2598268-2598268 on Build GRCh38
rs956707820 Chr.16: 2598170-2598170 on Build GRCh38
rs1048097481 Chr.16: 2598121-2598121 on Build GRCh38
rs375417064 Chr.16: 2598307-2598307 on Build GRCh38
rs951493040 Chr.16: 2598420-2598422 on Build GRCh38
rs906349928 Chr.16: 2598342-2598342 on Build GRCh38
rs995184591 Chr.16: 2597999-2597999 on Build GRCh38
rs547002823 Chr.16: 2598276-2598276 on Build GRCh38
rs189787811 Chr.16: 2598176-2598176 on Build GRCh38
rs780858375 Chr.16: 2598377-2598377 on Build GRCh38
rs941599686 Chr.16: 2598445-2598445 on Build GRCh38
rs958033407 Chr.16: 2598256-2598256 on Build GRCh38
rs185952911 Chr.16: 2598394-2598394 on Build GRCh38
rs559332782 Chr.16: 2597917-2597917 on Build GRCh38
rs976811876 Chr.16: 2598398-2598398 on Build GRCh38
rs113332295 Chr.16: 2598272-2598272 on Build GRCh38
rs949548322 Chr.16: 2598093-2598093 on Build GRCh38
rs894975750 Chr.16: 2598209-2598209 on Build GRCh38
rs932391917 Chr.16: 2598275-2598275 on Build GRCh38
rs535850233 Chr.16: 2598347-2598347 on Build GRCh38
rs1037770118 Chr.16: 2598325-2598325 on Build GRCh38
rs1040790639 Chr.16: 2598130-2598130 on Build GRCh38
rs776687154 Chr.16: 2597954-2597954 on Build GRCh38
rs181078294 Chr.16: 2598262-2598262 on Build GRCh38
rs935396654 Chr.16: 2597953-2597953 on Build GRCh38
rs922779617 Chr.16: 2598404-2598404 on Build GRCh38
rs1049958289 Chr.16: 2598427-2598427 on Build GRCh38
rs953423940 Chr.16: 2597921-2597921 on Build GRCh38
rs886896054 Chr.16: 2598017-2598017 on Build GRCh38
rs796987567 Chr.16: 2598246-2598247 on Build GRCh38
rs891675778 Chr.16: 2598303-2598303 on Build GRCh38
rs752529981 Chr.16: 2598006-2598006 on Build GRCh38
rs371038846 Chr.16: 2598105-2598105 on Build GRCh38
rs999408916 Chr.16: 2598355-2598355 on Build GRCh38
rs552190173 Chr.16: 2598286-2598287 on Build GRCh38
rs984765391 Chr.16: 2598249-2598249 on Build GRCh38
rs1047281541 Chr.16: 2598285-2598285 on Build GRCh38
rs953005979 Chr.16: 2598345-2598345 on Build GRCh38
rs989554225 Chr.16: 2598067-2598067 on Build GRCh38
rs750481961 Chr.16: 2598386-2598386 on Build GRCh38
rs1045351459 Chr.16: 2597983-2597983 on Build GRCh38
rs1012074079 Chr.16: 2598245-2598245 on Build GRCh38
rs961600586 Chr.16: 2598381-2598381 on Build GRCh38
rs926780855 Chr.16: 2598190-2598190 on Build GRCh38
rs980996213 Chr.16: 2598180-2598180 on Build GRCh38
rs979803400 Chr.16: 2597929-2597929 on Build GRCh38
rs571210384 Chr.16: 2598252-2598252 on Build GRCh38
rs925705174 Chr.16: 2598431-2598431 on Build GRCh38
rs1009576541 Chr.16: 2598306-2598306 on Build GRCh38
rs762295195 Chr.16: 2597968-2597968 on Build GRCh38
rs753864539 Chr.16: 2598212-2598212 on Build GRCh38
rs374470586 Chr.16: 2598073-2598073 on Build GRCh38
rs1002680332 Chr.16: 2598336-2598336 on Build GRCh38
rs904479944 Chr.16: 2598152-2598152 on Build GRCh38
rs559150292 Chr.16: 2598413-2598413 on Build GRCh38
rs886082649 Chr.16: 2598165-2598165 on Build GRCh38
rs970839400 Chr.16: 2598335-2598335 on Build GRCh38
rs983442247 Chr.16: 2598423-2598423 on Build GRCh38
rs1028567898 Chr.16: 2598149-2598149 on Build GRCh38
rs3087784 Chr.16: 2597912-2597912 on Build GRCh38
rs984266478 Chr.16: 2598351-2598351 on Build GRCh38
rs561226269 Chr.16: 2598050-2598050 on Build GRCh38
rs900999417 Chr.16: 2598033-2598033 on Build GRCh38
rs754005480 Chr.16: 2598197-2598197 on Build GRCh38
rs964150763 Chr.16: 2598395-2598395 on Build GRCh38
rs911269169 Chr.16: 2598271-2598271 on Build GRCh38
rs1003914338 Chr.16: 2598018-2598018 on Build GRCh38
rs543128805 Chr.16: 2598138-2598138 on Build GRCh38
rs908807426 Chr.16: 2598364-2598364 on Build GRCh38
rs572774850 Chr.16: 2598003-2598003 on Build GRCh38
rs1052903909 Chr.16: 2598460-2598460 on Build GRCh38
rs905417586 Chr.16: 2597988-2597988 on Build GRCh38
rs529226935 Chr.16: 2598036-2598036 on Build GRCh38
rs904342906 Chr.16: 2598278-2598278 on Build GRCh38
rs907542314 Chr.16: 2598299-2598299 on Build GRCh38
rs978098029 Chr.16: 2598039-2598039 on Build GRCh38
rs997224730 Chr.16: 2597935-2597935 on Build GRCh38
rs369818229 Chr.16: 2598070-2598070 on Build GRCh38
rs560922224 Chr.16: 2598403-2598403 on Build GRCh38
rs1030883247 Chr.16: 2598357-2598357 on Build GRCh38
rs375635017 Chr.16: 2597952-2597952 on Build GRCh38
rs528250860 Chr.16: 2598167-2598167 on Build GRCh38
rs939801240 Chr.16: 2598126-2598126 on Build GRCh38
rs572787105 Chr.16: 2598415-2598415 on Build GRCh38
rs959207591 Chr.16: 2598005-2598005 on Build GRCh38
rs563682651 Chr.16: 2597927-2597927 on Build GRCh38
rs148121766 Chr.16: 2598043-2598043 on Build GRCh38
rs374612917 Chr.16: 2598267-2598268 on Build GRCh38
rs1032374700 Chr.16: 2598164-2598164 on Build GRCh38
rs988114208 Chr.16: 2598171-2598171 on Build GRCh38
rs970765798 Chr.16: 2598179-2598179 on Build GRCh38
rs751303008 Chr.16: 2598297-2598297 on Build GRCh38
rs13335284 Chr.16: 2598069-2598069 on Build GRCh38
rs1033880199 Chr.16: 2598344-2598344 on Build GRCh38
rs539284441 Chr.16: 2598399-2598399 on Build GRCh38
rs886756377 Chr.16: 2598122-2598122 on Build GRCh38
rs981306719 Chr.16: 2598095-2598095 on Build GRCh38
rs996643932 Chr.16: 2598144-2598144 on Build GRCh38
rs563611183 Chr.16: 2598131-2598131 on Build GRCh38
rs969007414 Chr.16: 2598045-2598045 on Build GRCh38
rs1000504179 Chr.16: 2598158-2598158 on Build GRCh38
rs1040654421 Chr.16: 2598260-2598260 on Build GRCh38
rs559554679 Chr.16: 2598283-2598283 on Build GRCh38

View Full Product Details