Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 181241981-181242473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RACK1
Entrez Gene ID
10399
Gene Name
receptor for activated C kinase 1
Gene Aliases
GNB2L1, Gnb2-rs1, H12.3, HLC-7, PIG21
UniGene
Hs.5662
Gene Chromosome Location
Chr.5: 181236928-181243906 on Build GRCh38
Transcripts
NM_006098.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5194995 c.260T>G p.L87R Chr.5: 181242195-181242195 on Build GRCh38
6672306 c.224G>A p.G75D Chr.5: 181242231-181242231 on Build GRCh38
5967108 c.169C>G p.R57G Chr.5: 181242286-181242286 on Build GRCh38
1208410 c.281C>T p.T94M Chr.5: 181242174-181242174 on Build GRCh38
3013136 c.240A>G p.S80S Chr.5: 181242215-181242215 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754741658 Chr.5: 181242354-181242354 on Build GRCh38
rs768257936 Chr.5: 181242151-181242151 on Build GRCh38
rs919140594 Chr.5: 181242120-181242120 on Build GRCh38
rs11540190 Chr.5: 181242266-181242266 on Build GRCh38
rs981259957 Chr.5: 181242140-181242140 on Build GRCh38
rs879143243 Chr.5: 181242172-181242172 on Build GRCh38
rs1022809028 Chr.5: 181242107-181242107 on Build GRCh38
rs965668700 Chr.5: 181242028-181242028 on Build GRCh38
rs990438592 Chr.5: 181242098-181242098 on Build GRCh38
rs17657699 Chr.5: 181242136-181242136 on Build GRCh38
rs781561432 Chr.5: 181242040-181242041 on Build GRCh38
rs753979165 Chr.5: 181242292-181242292 on Build GRCh38
rs985869661 Chr.5: 181242050-181242050 on Build GRCh38
rs534105679 Chr.5: 181242277-181242277 on Build GRCh38
rs771622332 Chr.5: 181242191-181242191 on Build GRCh38
rs928069129 Chr.5: 181242269-181242269 on Build GRCh38
rs1020971589 Chr.5: 181242362-181242363 on Build GRCh38
rs889256648 Chr.5: 181242034-181242034 on Build GRCh38
rs369147803 Chr.5: 181242139-181242139 on Build GRCh38
rs921432255 Chr.5: 181242039-181242039 on Build GRCh38
rs59085681 Chr.5: 181242275-181242275 on Build GRCh38
rs11540188 Chr.5: 181242181-181242181 on Build GRCh38
rs765639929 Chr.5: 181242087-181242087 on Build GRCh38
rs368192532 Chr.5: 181242338-181242338 on Build GRCh38
rs371464588 Chr.5: 181242305-181242305 on Build GRCh38
rs192722247 Chr.5: 181242348-181242348 on Build GRCh38
rs775993596 Chr.5: 181242376-181242376 on Build GRCh38
rs773060507 Chr.5: 181242069-181242069 on Build GRCh38
rs200614777 Chr.5: 181242377-181242377 on Build GRCh38
rs767259927 Chr.5: 181242055-181242055 on Build GRCh38
rs757768792 Chr.5: 181242361-181242361 on Build GRCh38
rs754630422 Chr.5: 181242389-181242389 on Build GRCh38
rs11540177 Chr.5: 181242294-181242294 on Build GRCh38
rs776627849 Chr.5: 181242392-181242392 on Build GRCh38
rs995460380 Chr.5: 181242021-181242021 on Build GRCh38
rs554824522 Chr.5: 181242159-181242159 on Build GRCh38
rs747670943 Chr.5: 181242359-181242359 on Build GRCh38
rs1002028942 Chr.5: 181242282-181242282 on Build GRCh38
rs1036058407 Chr.5: 181242109-181242109 on Build GRCh38
rs34318897 Chr.5: 181242186-181242187 on Build GRCh38
rs893916495 Chr.5: 181242337-181242337 on Build GRCh38
rs11540172 Chr.5: 181242235-181242235 on Build GRCh38
rs958272407 Chr.5: 181242014-181242014 on Build GRCh38
rs11249777 Chr.5: 181242239-181242239 on Build GRCh38
rs11540166 Chr.5: 181242237-181242237 on Build GRCh38
rs11540167 Chr.5: 181242295-181242295 on Build GRCh38
rs889355442 Chr.5: 181242452-181242452 on Build GRCh38
rs58171665 Chr.5: 181242441-181242443 on Build GRCh38
rs750193394 Chr.5: 181242333-181242333 on Build GRCh38
rs1031295546 Chr.5: 181242081-181242081 on Build GRCh38
rs764157587 Chr.5: 181242281-181242281 on Build GRCh38
rs2287715 Chr.5: 181242370-181242370 on Build GRCh38
rs780890063 Chr.5: 181242154-181242154 on Build GRCh38
rs753211416 Chr.5: 181242430-181242430 on Build GRCh38
rs78732512 Chr.5: 181242076-181242076 on Build GRCh38
rs574279434 Chr.5: 181242366-181242366 on Build GRCh38
rs758004528 Chr.5: 181242121-181242121 on Build GRCh38
rs772297666 Chr.5: 181242373-181242373 on Build GRCh38
rs761914837 Chr.5: 181242393-181242393 on Build GRCh38
rs145195805 Chr.5: 181242024-181242024 on Build GRCh38
rs976892737 Chr.5: 181242033-181242033 on Build GRCh38
rs200609705 Chr.5: 181242448-181242450 on Build GRCh38
rs559260027 Chr.5: 181242385-181242385 on Build GRCh38
rs1049756177 Chr.5: 181242434-181242434 on Build GRCh38
rs759838218 Chr.5: 181242349-181242349 on Build GRCh38
rs748813655 Chr.5: 181242367-181242370 on Build GRCh38
rs1009611944 Chr.5: 181242017-181242017 on Build GRCh38
rs149150120 Chr.5: 181242043-181242043 on Build GRCh38
rs758151181 Chr.5: 181242335-181242335 on Build GRCh38
rs765093139 Chr.5: 181242323-181242323 on Build GRCh38
rs759974310 Chr.5: 181242071-181242071 on Build GRCh38
rs113616643 Chr.5: 181242171-181242171 on Build GRCh38
rs772294196 Chr.5: 181242272-181242272 on Build GRCh38
rs749806666 Chr.5: 181242174-181242174 on Build GRCh38
rs11540192 Chr.5: 181242262-181242262 on Build GRCh38
rs146525906 Chr.5: 181242453-181242453 on Build GRCh38
rs796760425 Chr.5: 181242447-181242449 on Build GRCh38
rs759820556 Chr.5: 181242271-181242271 on Build GRCh38

View Full Product Details