Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYPL2
Entrez Gene ID
284612
Gene Name
synaptophysin like 2
Gene Aliases
MG29
Chromosome Location
Chr.1: 109466478 - 109482142 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040709.1 NP_001035799.1 2 - 3 496 58
XM_011541283.2 XP_011539585.1 2 - 3 496 58
XM_011541284.2 XP_011539586.1 2 - 3 496 58
XM_011541285.1 XP_011539587.1 2 - 3 496 58
GenBank mRNA AB158472.1 - 2 - 3 130 58
AI983207.1 - 2 - 3 349 58
AK131459.1 - 2 - 3 496 58
AK289513.1 - 2 - 3 430 58
AK293082.1 - 2 - 3 372 58
AK302566.1 - 2 - 3 416 58
DA097637.1 - 2 - 3 430 58
DB071160.1 - 2 - 3 496 58
DB291596.1 - 2 - 3 416 58

View Full Product Details