Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MEIS1
Entrez Gene ID
4211
Gene Name
Meis homeobox 1
Gene Aliases
-
Chromosome Location
Chr.2: 66435125 - 66572765 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002398.2 NP_002389.1 1 - 2 466 111
XM_005264322.2 XP_005264379.1 1 - 2 741 111
XM_017004142.1 XP_016859631.1 1 - 2 741 111
XM_017004145.1 XP_016859634.1 1 - 2 741 111
XM_017004146.1 XP_016859635.1 1 - 2 741 111
GenBank mRNA AK314949.1 - 1 - 2 216 111
AL832770.1 - 1 - 2 479 111
BC043503.1 - 1 - 2 461 111
U85707.1 - 1 - 2 74 111

View Full Product Details