Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FEM1A
Entrez Gene ID
55527
Gene Name
fem-1 homolog A
Gene Aliases
EPRAP
Chromosome Location
Chr.19: 4791716 - 4795559 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018708.2 NP_061178.1 1 - 1 1907 80
GenBank mRNA AJ311359.1 - 1 - 1 1767 80
AK074232.1 - 1 - 1 1865 80
AK126532.1 - 2 - 2 1635 80
AK315554.1 - 1 - 1 1879 80
AL359589.1 - 1 - 1 45 80
AL365408.1 - 1 - 1 362 80
AY356352.1 - 1 - 1 1861 80
BC004988.1 - 1 - 1 1865 80
CR749732.1 - 1 - 1 45 80

View Full Product Details