The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031908.4 | NP_114114.2 | 2 - 3 | 385 | 88 |
XM_011534425.2 | XP_011532727.1 | 3 - 4 | 517 | 88 | |
GenBank mRNA | AF329836.1 | - | 2 - 3 | 393 | 88 |
AY358839.1 | - | 2 - 3 | 385 | 88 | |
BC011699.2 | - | 2 - 3 | 366 | 88 | |
BC054506.1 | - | 2 - 3 | 359 | 88 |