The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138289.3 | NP_612146.1 | 1 - 1 | 1399 | 69 |
GenBank mRNA | BC014597.1 | - | 1 - 1 | 1397 | 69 |
DB510229.1 | - | 1 - 1 | 380 | 69 |