Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ELOVL3
Entrez Gene ID
83401
Gene Name
ELOVL fatty acid elongase 3
Gene Aliases
CIG-30, CIG30
Chromosome Location
Chr.10: 102224767 - 102229589 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152310.2 NP_689523.1 3 - 4 653 64
XM_011540245.2 XP_011538547.1 4 - 5 1643 64
GenBank mRNA BC034344.1 - 3 - 4 545 64

View Full Product Details