Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FGD5
Entrez Gene ID
152273
Gene Name
FYVE, RhoGEF and PH domain containing 5
Gene Aliases
ZFYVE23
Chromosome Location
Chr.3: 14818962 - 14934565 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320276.1 NP_001307205.1 9 - 10 3376 58
NM_152536.3 NP_689749.3 9 - 10 3376 58
XM_011533422.1 XP_011531724.1 9 - 10 3376 58
XM_011533423.1 XP_011531725.1 9 - 10 3376 58
GenBank mRNA AK097276.1 - 9 - 10 1889 58
AK123054.1 - 6 - 7 568 58
AK131078.1 - 9 - 10 3040 58
BC035364.1 - 6 - 7 813 58
BC132814.1 - 9 - 10 3014 58
BC132816.1 - 9 - 10 3014 58
BC144300.1 - 9 - 10 3014 58
BX538312.1 - 9 - 10 1517 58
BX640820.1 - 9 - 10 2317 58

View Full Product Details