The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320276.1 | NP_001307205.1 | 12 - 13 | 3602 | 71 |
NM_152536.3 | NP_689749.3 | 12 - 13 | 3602 | 71 | |
XM_011533422.1 | XP_011531724.1 | 12 - 13 | 3602 | 71 | |
XM_011533423.1 | XP_011531725.1 | 12 - 13 | 3602 | 71 | |
GenBank mRNA | AK096856.1 | - | 1 - 2 | 92 | 71 |
AK097276.1 | - | 12 - 13 | 2115 | 71 | |
AK123054.1 | - | 9 - 10 | 794 | 71 | |
AK131078.1 | - | 12 - 13 | 3266 | 71 | |
BC035364.1 | - | 9 - 10 | 1039 | 71 | |
BC132814.1 | - | 12 - 13 | 3240 | 71 | |
BC132816.1 | - | 12 - 13 | 3240 | 71 | |
BC144300.1 | - | 12 - 13 | 3240 | 71 | |
BX538312.1 | - | 12 - 13 | 1743 | 71 | |
BX640820.1 | - | 12 - 13 | 2543 | 71 |