Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FGD5
Entrez Gene ID
152273
Gene Name
FYVE, RhoGEF and PH domain containing 5
Gene Aliases
ZFYVE23
Chromosome Location
Chr.3: 14818962 - 14934565 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320276.1 NP_001307205.1 14 - 15 3775 66
NM_152536.3 NP_689749.3 14 - 15 3775 66
XM_011533422.1 XP_011531724.1 14 - 15 3775 66
XM_011533423.1 XP_011531725.1 14 - 15 3775 66
GenBank mRNA AK096856.1 - 3 - 4 265 66
AK097276.1 - 14 - 15 2288 66
AK123054.1 - 11 - 12 967 66
AK131078.1 - 14 - 15 3439 66
BC035364.1 - 11 - 12 1212 66
BC132814.1 - 14 - 15 3413 66
BC132816.1 - 14 - 15 3413 66
BC144300.1 - 14 - 15 3413 66
BX538312.1 - 14 - 15 1916 66
BX640820.1 - 14 - 15 2716 66

View Full Product Details