Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FGD5
Entrez Gene ID
152273
Gene Name
FYVE, RhoGEF and PH domain containing 5
Gene Aliases
ZFYVE23
Chromosome Location
Chr.3: 14818962 - 14934565 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320276.1 NP_001307205.1 15 - 16 3913 122
NM_152536.3 NP_689749.3 15 - 16 3913 122
XM_011533422.1 XP_011531724.1 15 - 16 3913 122
XM_011533423.1 XP_011531725.1 15 - 16 3913 122
GenBank mRNA AK096856.1 - 4 - 5 403 122
AK097276.1 - 15 - 16 2426 122
AK123054.1 - 12 - 13 1105 122
AK131078.1 - 15 - 16 3577 122
BC035364.1 - 12 - 13 1350 122
BC132814.1 - 15 - 16 3551 122
BC132816.1 - 15 - 16 3551 122
BC144300.1 - 15 - 16 3551 122
BX538312.1 - 15 - 16 2054 122

View Full Product Details