The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152890.5 | NP_690850.2 | 26 - 27 | 2712 | 95 |
XM_017000924.1 | XP_016856413.1 | 28 - 29 | 2885 | 95 | |
XM_017000925.1 | XP_016856414.1 | 28 - 29 | 2840 | 95 | |
XM_017000926.1 | XP_016856415.1 | 27 - 28 | 3082 | 95 | |
XM_017000927.1 | XP_016856416.1 | 27 - 28 | 3082 | 95 | |
XM_017000928.1 | XP_016856417.1 | 9 - 10 | 1794 | 95 | |
XM_017000929.1 | XP_016856418.1 | 8 - 9 | 1748 | 95 | |
GenBank mRNA | AB209686.1 | - | 3 - 4 | 2104 | 95 |
AY244357.1 | - | 26 - 27 | 2702 | 95 | |
BC113654.1 | - | 26 - 27 | 2709 | 95 | |
BC117170.1 | - | 26 - 27 | 2709 | 95 | |
BC143715.1 | - | 27 - 28 | 2735 | 95 | |
BC143717.1 | - | 26 - 27 | 2705 | 95 | |
BC143718.1 | - | 27 - 28 | 2811 | 95 |