The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152890.5 | NP_690850.2 | 40 - 41 | 3558 | 85 |
XM_017000924.1 | XP_016856413.1 | 42 - 43 | 3731 | 85 | |
XM_017000925.1 | XP_016856414.1 | 42 - 43 | 3686 | 85 | |
XM_017000926.1 | XP_016856415.1 | 41 - 42 | 3928 | 85 | |
XM_017000927.1 | XP_016856416.1 | 40 - 41 | 3874 | 85 | |
XM_017000928.1 | XP_016856417.1 | 23 - 24 | 2640 | 85 | |
XM_017000929.1 | XP_016856418.1 | 22 - 23 | 2594 | 85 | |
GenBank mRNA | AB209686.1 | - | 17 - 18 | 2950 | 85 |
AY244357.1 | - | 40 - 41 | 3548 | 85 | |
BC113654.1 | - | 40 - 41 | 3555 | 85 | |
BC117170.1 | - | 40 - 41 | 3555 | 85 | |
BC143715.1 | - | 40 - 41 | 3527 | 85 | |
BC143717.1 | - | 40 - 41 | 3551 | 85 | |
BC143718.1 | - | 40 - 41 | 3603 | 85 |