The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153339.1 | NP_699170.1 | 3 - 4 | 355 | 86 |
XM_005244720.3 | XP_005244777.1 | 3 - 4 | 407 | 86 | |
GenBank mRNA | AK027721.1 | - | 1 - 2 | 342 | 86 |
AK075292.1 | - | 3 - 4 | 372 | 86 | |
AK298218.1 | - | 2 - 3 | 265 | 86 | |
BC034304.1 | - | 3 - 4 | 355 | 86 |