The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153444.1 | NP_703145.1 | 1 - 1 | 453 | 110 |
GenBank mRNA | AF173006.1 | - | 1 - 1 | 453 | 110 |
AF220494.1 | - | 1 - 1 | 114 | 110 | |
BC101710.1 | - | 1 - 1 | 484 | 110 | |
BC113557.1 | - | 1 - 1 | 484 | 110 |